Product Details

SNP ID
rs140136558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45255338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTCTCGCTCCTCCACAGTGG[C/T]CAGGAGGGGCTCTGTGGAAGGCTCC
Phenotype
MIM: 604656 MIM: 604655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMNL1 PubMed Links

Gene Details

Gene
FMNL1
Gene Name
formin like 1
There are no transcripts associated with this gene.

Gene
MAP3K14
Gene Name
mitogen-activated protein kinase kinase kinase 14
There are no transcripts associated with this gene.

Gene
MAP3K14-AS1
Gene Name
MAP3K14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SPATA32
Gene Name
spermatogenesis associated 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152343.2 940 Missense Mutation ACC,GCC T282A NP_689556.2

View Full Product Details