Product Details

SNP ID
rs140180116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50063973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGAGGGCAGGAGCTGGGGGTC[G/T]CCTGGCACCAAGAATCTGTAAATGT
Phenotype
MIM: 605025
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGA3 PubMed Links

Gene Details

Gene
ITGA3
Gene Name
integrin subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002204.3 Intron NP_002195.1
XM_005257308.2 Intron XP_005257365.1
Gene
LOC284080
Gene Name
uncharacterized LOC284080
There are no transcripts associated with this gene.

View Full Product Details