Product Details

SNP ID
rs140183637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47655754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTATAACAGCAACAATGAGCCTG[A/G]AAGATGAGTTTTGTCCCCATCTCTC
Phenotype
MIM: 602738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPNB1 PubMed Links

Gene Details

Gene
KPNB1
Gene Name
karyopherin subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276453.1 Intron NP_001263382.1
NM_002265.5 Intron NP_002256.2

View Full Product Details