Product Details

SNP ID
rs140202481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32444573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGAGAGCCCAGTCTCTACTCA[A/G]CACCGACCGGGAGGCCTCCATCGAC
Phenotype
MIM: 604449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMD11 PubMed Links

Gene Details

Gene
PSMD11
Gene Name
proteasome 26S subunit, non-ATPase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270482.1 111 Missense Mutation AAC,AGC N17S NP_001257411.1
NM_002815.3 111 Missense Mutation AAC,AGC N17S NP_002806.2

View Full Product Details