Product Details

SNP ID
rs140252396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7337313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCATGACCCCCTCTTTCCCAGA[A/G]CCTCTATTGAGCTGGTGGAAGCCGA
Phenotype
MIM: 607763 MIM: 615865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 261 Missense Mutation ACC,GCC T19A NP_055531.1
Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
There are no transcripts associated with this gene.

View Full Product Details