Product Details

SNP ID
rs140332327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58192922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCCTGTGAGGGCACTGACCCA[A/G]GTAATAGTCCCCTAGACAGGCAAGG
Phenotype
MIM: 131399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPX PubMed Links

Gene Details

Gene
EPX
Gene Name
eosinophil peroxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000502.5 199 Missense Mutation ACC,GCC T26A NP_000493.1

View Full Product Details