Product Details

SNP ID
rs140424050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28381340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAGGAGACATGCCAGAGGCTGG[A/T]GGCGGCCGGCGGCCTGGACGCGGTG
Phenotype
MIM: 607732
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SARM1 PubMed Links

Gene Details

Gene
SARM1
Gene Name
sterile alpha and TIR motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015077.3 979 Missense Mutation GAG,GTG E203V NP_055892.2

View Full Product Details