Product Details

SNP ID
rs140627736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81955469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAGCCGCAGGCCCTCCTGCACC[A/G]GCTGCCCCGTCAGGTGCACGTTGTC
Phenotype
MIM: 609847
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYADML2 PubMed Links

Gene Details

Gene
MYADML2
Gene Name
myeloid associated differentiation marker like 2
There are no transcripts associated with this gene.

Gene
NOTUM
Gene Name
NOTUM, palmitoleoyl-protein carboxylesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178493.5 1336 Missense Mutation CCG,CTG P355L NP_848588.3

View Full Product Details