Product Details

SNP ID
rs140648663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44804696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCTTTTGTTGGACCCAGCTTT[A/G]GACCCCACTTTGGCCTTCTTCTCCC
Phenotype
MIM: 608655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJC1 PubMed Links

Gene Details

Gene
GJC1
Gene Name
gap junction protein gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080383.1 1390 Silent Mutation TCC,TCT S374S NP_001073852.1
NM_005497.3 1390 Silent Mutation TCC,TCT S374S NP_005488.2
XM_005256920.1 1390 Silent Mutation TCC,TCT S374S XP_005256977.1
XM_005256921.1 1390 Silent Mutation TCC,TCT S374S XP_005256978.1

View Full Product Details