Product Details

SNP ID
rs140866344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78797150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTCCACCTCAGTGAAGACCTG[A/C]GGGTTATTGCAGCTGCAAGGGAAGC
Phenotype
MIM: 612543
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
USP36 PubMed Links

Gene Details

Gene
USP36
Gene Name
ubiquitin specific peptidase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321291.1 4387 UTR 3 NP_001308220.1
XM_005257542.2 4387 UTR 3 XP_005257599.1
XM_005257543.2 4387 UTR 3 XP_005257600.1
XM_011525065.1 4387 UTR 3 XP_011523367.1
XM_011525066.2 4387 UTR 3 XP_011523368.1
XM_011525067.2 4387 Intron XP_011523369.1
XM_011525068.1 4387 UTR 3 XP_011523370.1
XM_011525069.2 4387 UTR 3 XP_011523371.1
XM_011525070.1 4387 UTR 3 XP_011523372.1
XM_011525071.2 4387 Intron XP_011523373.1
XM_017024900.1 4387 Intron XP_016880389.1
XM_017024901.1 4387 UTR 3 XP_016880390.1

View Full Product Details