Product Details

SNP ID
rs141035851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27472245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTGGGGACATTGTGCTCCTCAT[A/G]GGTTTCTTCCCGTAGGATGGGGCAA
Phenotype
MIM: 601132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KSR1 PubMed Links

Gene Details

Gene
KSR1
Gene Name
kinase suppressor of ras 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014238.1 Intron NP_055053.1
XM_006722154.3 Intron XP_006722217.1
XM_011525429.2 Intron XP_011523731.1
XM_011525430.2 Intron XP_011523732.1
XM_011525431.2 Intron XP_011523733.1
XM_017025267.1 Intron XP_016880756.1
XM_017025268.1 Intron XP_016880757.1
XM_017025269.1 Intron XP_016880758.1
XM_017025270.1 Intron XP_016880759.1
XM_017025271.1 Intron XP_016880760.1
XM_017025272.1 Intron XP_016880761.1
XM_017025273.1 Intron XP_016880762.1
XM_017025274.1 Intron XP_016880763.1
XM_017025275.1 Intron XP_016880764.1
XM_017025276.1 Intron XP_016880765.1
XM_017025277.1 Intron XP_016880766.1
XM_017025278.1 Intron XP_016880767.1
XM_017025279.1 Intron XP_016880768.1
XM_017025280.1 Intron XP_016880769.1

View Full Product Details