Product Details

SNP ID
rs141144435
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43953131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCGCGACCTGACGGGGCGGTCCT[C/T]GCCGTCGGGGAAGAACGTTTTGGAA
Phenotype
MIM: 167780 MIM: 600781
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PPY PubMed Links
Additional Information
For this assay, SNP(s) [rs162430] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PPY
Gene Name
pancreatic polypeptide
There are no transcripts associated with this gene.

Gene
PYY
Gene Name
peptide YY
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004160.5 1398 Intron NP_004151.3
XM_011525035.1 1398 Missense Mutation AAG,GAG K83E XP_011523337.1

View Full Product Details