Product Details

SNP ID
rs141190899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44761557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGATTCCAGATACTTCTGAGGG[A/G]TGTCTGAGAGCCTGCGGGGAGGGAA
Phenotype
MIM: 155120 MIM: 611661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM11 PubMed Links

Gene Details

Gene
ADAM11
Gene Name
ADAM metallopeptidase domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318933.1 Intron NP_001305862.1
NM_002390.5 Intron NP_002381.2
XM_005257373.3 Intron XP_005257430.1
XM_017024647.1 Intron XP_016880136.1
Gene
DBF4B
Gene Name
DBF4 zinc finger B
There are no transcripts associated with this gene.

View Full Product Details