Product Details

SNP ID
rs141254931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4960486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCTCCTCTCGATCTTGTTCATT[A/G]GCTGAAACTTTTTCTTGATCTGCCC
Phenotype
MIM: 611508 MIM: 131370 MIM: 610056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMTA2 PubMed Links

Gene Details

Gene
CAMTA2
Gene Name
calmodulin binding transcription activator 2
There are no transcripts associated with this gene.

Gene
ENO3
Gene Name
enolase 3
There are no transcripts associated with this gene.

Gene
MIR6864
Gene Name
microRNA 6864
There are no transcripts associated with this gene.

Gene
MIR6865
Gene Name
microRNA 6865
There are no transcripts associated with this gene.

Gene
SPAG7
Gene Name
sperm associated antigen 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004890.2 873 Missense Mutation CCA,CTA P72L NP_004881.2
XM_006721600.2 873 Missense Mutation CCA,CTA P21L XP_006721663.1

View Full Product Details