Product Details

SNP ID
rs141296055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81844087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTCTGCTTCTTCCAGGTCCTC[A/G]AGATCCTGGGATACAGGAAAAGGGG
Phenotype
MIM: 176790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
P4HB PubMed Links

Gene Details

Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000918.3 1649 Silent Mutation CTC,CTT L484L NP_000909.2
Gene
PPP1R27
Gene Name
protein phosphatase 1 regulatory subunit 27
There are no transcripts associated with this gene.

View Full Product Details