Product Details

SNP ID
rs141528543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82322210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGTTGGGGGGACAGCAGCTCC[A/C]GGGCTCCAAGTGGTGAGGGTTTGAA
Phenotype
MIM: 186820 MIM: 602602
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD7 PubMed Links

Gene Details

Gene
CD7
Gene Name
CD7 molecule
There are no transcripts associated with this gene.

Gene
SECTM1
Gene Name
secreted and transmembrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003004.2 1127 Missense Mutation CGG,CTG R233L NP_002995.1
XM_005256392.3 1127 Missense Mutation CGG,CTG R233L XP_005256449.1
XM_011523588.2 1127 Missense Mutation CGG,CTG R233L XP_011521890.1

View Full Product Details