Product Details

SNP ID
rs141547861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42669996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCAGGGCCAGGTAGGCGGCCAT[C/G]GCCTCAGCTCGGCCCATGCCCCGTA
Phenotype
MIM: 600240 MIM: 602346 MIM: 605785
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCR10 PubMed Links

Gene Details

Gene
CCR10
Gene Name
C-C motif chemokine receptor 10
There are no transcripts associated with this gene.

Gene
CNTNAP1
Gene Name
contactin associated protein 1
There are no transcripts associated with this gene.

Gene
PLEKHH3
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024927.4 2238 Silent Mutation GCC,GCG A645A NP_079203.3
XM_017025113.1 2238 Silent Mutation GCC,GCG A734A XP_016880602.1
XM_017025114.1 2238 Silent Mutation GCC,GCG A731A XP_016880603.1
XM_017025115.1 2238 Silent Mutation GCC,GCG A690A XP_016880604.1
XM_017025116.1 2238 Silent Mutation GCC,GCG A689A XP_016880605.1
XM_017025117.1 2238 Silent Mutation GCC,GCG A674A XP_016880606.1
XM_017025118.1 2238 Silent Mutation GCC,GCG A642A XP_016880607.1
Gene
TUBG2
Gene Name
tubulin gamma 2
There are no transcripts associated with this gene.

View Full Product Details