Product Details

SNP ID
rs141549690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31304908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATCTACCTGCCCAATTTTGATC[A/G]TTTAAAACTGGTTTCCTTAAGCATT
Phenotype
MIM: 158381 MIM: 613113 MIM: 164345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVI2B PubMed Links

Gene Details

Gene
EVI2B
Gene Name
ecotropic viral integration site 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006495.3 967 Silent Mutation AAC,AAT N234N NP_006486.3
XM_005257946.3 967 Silent Mutation AAC,AAT N249N XP_005258003.1
Gene
NF1
Gene Name
neurofibromin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000267.3 967 Intron NP_000258.1
NM_001042492.2 967 Intron NP_001035957.1
NM_001128147.2 967 Intron NP_001121619.1
XM_005257983.2 967 Intron XP_005258040.1
XM_005257984.2 967 Intron XP_005258041.1
XM_006721922.2 967 Intron XP_006721985.1
XM_006721924.2 967 Intron XP_006721987.1
XM_006721925.2 967 Intron XP_006721988.1
XM_006721926.3 967 Intron XP_006721989.1
XM_011524852.2 967 Intron XP_011523154.1
XM_011524857.2 967 Intron XP_011523159.1
XM_017024689.1 967 Intron XP_016880178.1
XM_017024690.1 967 Intron XP_016880179.1
Gene
OMG
Gene Name
oligodendrocyte myelin glycoprotein
There are no transcripts associated with this gene.

View Full Product Details