Product Details

SNP ID
rs141743623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36950259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGTTTGATGAAGGGGAAGATG[A/G]GGAAGGTGATTTCCTAGTAGTGGGT
Phenotype
MIM: 608463 MIM: 601999
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AATF PubMed Links

Gene Details

Gene
AATF
Gene Name
apoptosis antagonizing transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012138.3 388 Missense Mutation GAG,GGG E46G NP_036270.1
XM_011524611.1 388 Missense Mutation GAG,GGG E46G XP_011522913.1
Gene
LHX1
Gene Name
LIM homeobox 1
There are no transcripts associated with this gene.

View Full Product Details