Product Details

SNP ID
rs141836960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4809161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAGAGAGATGCCCTCTCTACCC[C/T]GGGCAGGTCCTGAGGGCTCCACCAG
Phenotype
MIM: 602384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107984973 PubMed Links

Gene Details

Gene
LOC107984973
Gene Name
atherin-like
There are no transcripts associated with this gene.

Gene
PLD2
Gene Name
phospholipase D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243108.1 520 Missense Mutation CGG,TGG R149W NP_001230037.1
NM_002663.4 520 Missense Mutation CGG,TGG R149W NP_002654.3
XM_005256696.2 520 Intron XP_005256753.1
XM_017024764.1 520 UTR 5 XP_016880253.1

View Full Product Details