Product Details

SNP ID
rs141837882
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60043953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAAAGTCTATGGTGTCTTCACT[C/G]TTCTGTAAAGCGGTGACCAATGCAT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HEATR6 PubMed Links

Gene Details

Gene
HEATR6
Gene Name
HEAT repeat containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022070.4 5110 Missense Mutation AAC,AAG N1052K NP_071353.4
XM_005257570.3 5110 Missense Mutation AAC,AAG N884K XP_005257627.1
XM_011525110.2 5110 Missense Mutation AAC,AAG N1016K XP_011523412.1
XM_011525111.2 5110 Missense Mutation AAC,AAG N1005K XP_011523413.1
XM_017024936.1 5110 Missense Mutation AAC,AAG N884K XP_016880425.1
XM_017024937.1 5110 Missense Mutation AAC,AAG N884K XP_016880426.1
XM_017024938.1 5110 Missense Mutation AAC,AAG N591K XP_016880427.1
XM_017024939.1 5110 Missense Mutation AAC,AAG N591K XP_016880428.1
Gene
MIR4737
Gene Name
microRNA 4737
There are no transcripts associated with this gene.

View Full Product Details