Product Details

SNP ID
rs141853377
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81899354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCATCAACACAGCTTCCCCAAC[A/G]CCTGGGCAGCACACCGGATCCCTGA
Phenotype
MIM: 604171 MIM: 614534 MIM: 607996 MIM: 602679
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links
Additional Information
For this assay, SNP(s) [rs61751632] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
There are no transcripts associated with this gene.

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002244.2 384 Silent Mutation ACA,ACG T74T NP_001002244.1
NM_001002245.2 384 Intron NP_001002245.1
NM_001002246.2 384 Intron NP_001002246.1
NM_001002247.2 384 Intron NP_001002247.1
NM_001002248.2 384 Intron NP_001002248.1
NM_001002249.2 384 Intron NP_001002249.1
NM_001289414.1 384 Intron NP_001276343.1
NM_001289415.1 384 Intron NP_001276344.1
NM_001289416.1 384 Intron NP_001276345.1
NM_001289417.1 384 Intron NP_001276346.1
NM_001289418.1 384 Intron NP_001276347.1
NM_001289419.1 384 Intron NP_001276348.1
NM_001289420.1 384 Intron NP_001276349.1
NM_016476.11 384 Intron NP_057560.8
Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

View Full Product Details