Product Details

SNP ID
rs141882639
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8096755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCTGGAAGGCGGCGATGCTCC[A/G]CCTCGGGGCCTCCTCTGTGAAGTGC
Phenotype
MIM: 603741 MIM: 607206
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ALOX12B PubMed Links

Gene Details

Gene
ALOX12B
Gene Name
arachidonate 12-lipoxygenase, 12R type
There are no transcripts associated with this gene.

Gene
ALOXE3
Gene Name
arachidonate lipoxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165960.1 2378 Missense Mutation CGG,TGG R802W NP_001159432.1
NM_021628.2 2378 Missense Mutation CGG,TGG R670W NP_067641.2
XM_017024921.1 2378 Intron XP_016880410.1
XM_017024922.1 2378 Intron XP_016880411.1
XM_017024923.1 2378 Intron XP_016880412.1
XM_017024924.1 2378 Intron XP_016880413.1
XM_017024925.1 2378 Intron XP_016880414.1
Gene
MIR4314
Gene Name
microRNA 4314
There are no transcripts associated with this gene.

View Full Product Details