Product Details
- SNP ID
-
rs142022888
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:76738940 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTAGTTTTATCTTCCACACAGCA[A/G]ACTGTCATCAGGAGCTTAAATAGGA
- Phenotype
-
MIM: 615262
MIM: 600813
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
METTL23
PubMed Links
Gene Details
- Gene
- METTL23
- Gene Name
- methyltransferase like 23
There are no transcripts associated with this gene.
- Gene
- MFSD11
- Gene Name
- major facilitator superfamily domain containing 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242532.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_001229461.1 |
NM_001242533.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_001229462.1 |
NM_001242534.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_001229463.1 |
NM_001242535.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_001229464.1 |
NM_001242536.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_001229465.1 |
NM_001242537.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_001229466.1 |
NM_024311.3 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
NP_077287.1 |
XM_011525236.2 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523538.1 |
XM_011525237.2 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523539.2 |
XM_011525238.2 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523540.2 |
XM_011525239.2 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523541.2 |
XM_011525240.2 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523542.2 |
XM_011525241.2 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523543.2 |
XM_011525242.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523544.1 |
XM_011525244.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_011523546.1 |
XM_011525247.2 |
835 |
UTR 5 |
|
|
XP_011523549.2 |
XM_017025065.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_016880554.1 |
XM_017025066.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_016880555.1 |
XM_017025067.1 |
835 |
Silent Mutation |
CAA,CAG |
Q33Q |
XP_016880556.1 |
XM_017025068.1 |
835 |
UTR 5 |
|
|
XP_016880557.1 |
XM_017025069.1 |
835 |
UTR 5 |
|
|
XP_016880558.1 |
XM_017025070.1 |
835 |
UTR 5 |
|
|
XP_016880559.1 |
XM_017025071.1 |
835 |
Intron |
|
|
XP_016880560.1 |
- Gene
- MIR636
- Gene Name
- microRNA 636
There are no transcripts associated with this gene.
- Gene
- SRSF2
- Gene Name
- serine and arginine rich splicing factor 2
There are no transcripts associated with this gene.
View Full Product Details