Product Details

SNP ID
rs142166462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36557116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCTTAAGCAACAGGATCTAAGT[A/G]TTGCCATGGTGGTGACATCACGCGA
Phenotype
MIM: 612275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGNBP2 PubMed Links

Gene Details

Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024835.4 263 Missense Mutation ATT,GTT I70V NP_079111.1
XM_005257689.2 263 Missense Mutation ATT,GTT I70V XP_005257746.1
XM_011525265.2 263 Missense Mutation ATT,GTT I39V XP_011523567.1
XM_011525266.1 263 Missense Mutation ATT,GTT I70V XP_011523568.1
XM_017025107.1 263 Missense Mutation ATT,GTT I39V XP_016880596.1

View Full Product Details