Product Details

SNP ID
rs142241491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5106637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGACAAACATTCGAAGAATATT[C/T]ACTGGATATTGCCAACAGTTATCTC
Phenotype
MIM: 604334 MIM: 616463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
USP6
Gene Name
ubiquitin specific peptidase 6
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 Intron NP_001307881.1
NM_001320953.1 Intron NP_001307882.1
NM_001320954.1 Intron NP_001307883.1
NM_001320955.1 Intron NP_001307884.1
NM_014519.3 Intron NP_055334.2
XM_011524006.2 Intron XP_011522308.1
XM_017025021.1 Intron XP_016880510.1
XM_017025022.1 Intron XP_016880511.1
XM_017025023.1 Intron XP_016880512.1

View Full Product Details