Product Details
- SNP ID
-
rs142246318
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:16417805 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCATGGAGTCACAGTTCCAGGGCG[A/G]GGACCGGAAATTCGCCCCTCAGATA
- Phenotype
-
MIM: 606676
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRPV2
PubMed Links
Gene Details
- Gene
- TRPV2
- Gene Name
- transient receptor potential cation channel subfamily V member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016113.4 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
NP_057197.2 |
XM_005256676.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_005256733.1 |
XM_005256677.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_005256734.1 |
XM_005256678.4 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_005256735.1 |
XM_006721541.3 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_006721604.1 |
XM_006721543.3 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_006721606.1 |
XM_011523922.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_011522224.1 |
XM_011523923.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_011522225.1 |
XM_011523925.2 |
536 |
Intron |
|
|
XP_011522227.1 |
XM_017024730.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_016880219.1 |
XM_017024731.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_016880220.1 |
XM_017024732.1 |
536 |
Missense Mutation |
GAG,GGG |
E46G |
XP_016880221.1 |
View Full Product Details