Product Details

SNP ID
rs142258568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69152392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCCTGACACCATCATGGCCATA[C/T]GGTCACACACAGCCTCAGCCTCTGA
Phenotype
MIM: 612508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCA10 PubMed Links

Gene Details

Gene
ABCA10
Gene Name
ATP binding cassette subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080282.3 5105 Missense Mutation NP_525021.3

View Full Product Details