Product Details

SNP ID
rs142359874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76040932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCTGCGGCCTGCAGGGAGCAC[A/T]TCTCTGACCGCACTTGAGTGATGAG
Phenotype
MIM: 604858
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SRP68 PubMed Links

Gene Details

Gene
SRP68
Gene Name
signal recognition particle 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260502.1 1606 Missense Mutation AAG,ATG K486M NP_001247431.1
NM_001260503.1 1606 Missense Mutation AAG,ATG K185M NP_001247432.1
NM_014230.3 1606 Missense Mutation AAG,ATG K524M NP_055045.2

View Full Product Details