Product Details

SNP ID
rs142522197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36013804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTAACTCTCCAGGAGTGAACAC[A/G]GGATGCTTCGTGGGGTGTAGGAGAT
Phenotype
MIM: 602494
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL23 PubMed Links

Gene Details

Gene
CCL23
Gene Name
C-C motif chemokine ligand 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005064.5 313 Missense Mutation CCG,CTG P81L NP_005055.3
NM_145898.3 313 Missense Mutation CCG,CTG P64L NP_665905.2

View Full Product Details