Product Details

SNP ID
rs142564218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43213349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTGAGTCTCCTTGGATGGGGT[C/T]AGATCTGCTCCCAGCTCTTGACCTC
Phenotype
MIM: 166945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00854 PubMed Links

Gene Details

Gene
LINC00854
Gene Name
long intergenic non-protein coding RNA 854
There are no transcripts associated with this gene.

Gene
NBR1
Gene Name
NBR1, autophagy cargo receptor
There are no transcripts associated with this gene.

Gene
TMEM106A
Gene Name
transmembrane protein 106A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291586.1 Intron NP_001278515.1
NM_001291587.1 Intron NP_001278516.1
NM_001291588.1 Intron NP_001278517.1
NM_145041.3 Intron NP_659478.1
XM_006721658.2 Intron XP_006721721.1
XM_006721659.3 Intron XP_006721722.1
XM_011524260.1 Intron XP_011522562.1
XM_017024113.1 Intron XP_016879602.1
XM_017024114.1 Intron XP_016879603.1

View Full Product Details