Product Details

SNP ID
rs142659640
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45255145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTTCCCTGCAGCAGAGGGGAC[A/G]TGGCTGGTGGCTGGAGAAGCTGGAT
Phenotype
MIM: 604656 MIM: 604655
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FMNL1 PubMed Links

Gene Details

Gene
FMNL1
Gene Name
formin like 1
There are no transcripts associated with this gene.

Gene
MAP3K14
Gene Name
mitogen-activated protein kinase kinase kinase 14
There are no transcripts associated with this gene.

Gene
MAP3K14-AS1
Gene Name
MAP3K14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SPATA32
Gene Name
spermatogenesis associated 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152343.2 1133 Missense Mutation ACG,ATG T346M NP_689556.2

View Full Product Details