Product Details

SNP ID
rs142895134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41422075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTGAAGGACACGTACCAAGGT[A/G]TGCTGGGCTTGGCGCTCCACCTCCA
Phenotype
MIM: 604541
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT37 PubMed Links

Gene Details

Gene
KRT37
Gene Name
keratin 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003770.4 1061 Silent Mutation CAC,CAT H338H NP_003761.3
Gene
LOC100505782
Gene Name
uncharacterized LOC100505782
There are no transcripts associated with this gene.

View Full Product Details