Product Details

SNP ID
rs142896224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5444376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAGGACAGACGACGGGTGGAT[A/G]GCCACTGGCTGGTGGGTGTCCGTGG
Phenotype
MIM: 601269 MIM: 614405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QBP PubMed Links

Gene Details

Gene
C1QBP
Gene Name
complement C1q binding protein
There are no transcripts associated with this gene.

Gene
DHX33
Gene Name
DEAH-box helicase 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199699.1 1993 Silent Mutation GCC,GCT A478A NP_001186628.1
NM_020162.3 1993 Silent Mutation GCC,GCT A651A NP_064547.2
XM_017024877.1 1993 Silent Mutation GCC,GCT A223A XP_016880366.1

View Full Product Details