Product Details

SNP ID
rs143088973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78855193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGGGCAGGGAGTGCAGAGGC[A/G]TGGAAGCTGCAGAAAACAAGGGCCA
Phenotype
MIM: 188825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIMP2 PubMed Links

Gene Details

Gene
TIMP2
Gene Name
TIMP metallopeptidase inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003255.4 1439 UTR 3 NP_003246.1

View Full Product Details