Product Details
- SNP ID
-
rs143089468
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:19243048 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTTTGGGCACTTTAATCACTGAG[C/T]TTTGGTGTCCTCATCTCTAAAGGGG
- Phenotype
-
MIM: 607263
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPN2
PubMed Links
Gene Details
- Gene
- EPN2
- Gene Name
- epsin 2
View Full Product Details