Product Details

SNP ID
rs143829372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48960890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAACACCACACTCCCCTACCTGA[C/G]CCTGCAGAGGTTTGTCTGATCCAGC
Phenotype
MIM: 137240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIP PubMed Links

Gene Details

Gene
GIP
Gene Name
gastric inhibitory polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004123.2 546 Missense Mutation CTC,GTC L150V NP_004114.1

View Full Product Details