Product Details

SNP ID
rs143841873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82362173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGTGCCCTCCGGTCAGCATGC[A/G]GTATGAGGAAGAGGGCATGTCCTAC
Phenotype
MIM: 615647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX19 PubMed Links

Gene Details

Gene
TEX19
Gene Name
testis expressed 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207459.3 420 Missense Mutation CAG,CGG Q8R NP_997342.1
XM_017024638.1 420 Missense Mutation CAG,CGG Q8R XP_016880127.1
XM_017024639.1 420 Missense Mutation CAG,CGG Q8R XP_016880128.1

View Full Product Details