Product Details

SNP ID
rs143844371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50278913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCAGACAGCCCAGTGGATTGC[C/T]GGGGGCCCCTGGAACTGCCCTCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM92 PubMed Links

Gene Details

Gene
TMEM92
Gene Name
transmembrane protein 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168215.1 393 Missense Mutation CGG,TGG R95W NP_001161687.1
NM_153229.2 393 Missense Mutation CGG,TGG R95W NP_694961.2
XM_006721730.3 393 Missense Mutation CGG,TGG R95W XP_006721793.1
Gene
TMEM92-AS1
Gene Name
TMEM92 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details