Product Details

SNP ID
rs143899860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39922670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCTGGTCGTACTTAGTGTAGAA[A/G]CTGGTGAGGAAGTACCTGGGAGGGG
Phenotype
MIM: 611221 MIM: 610075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMB PubMed Links

Gene Details

Gene
GSDMB
Gene Name
gasdermin B
There are no transcripts associated with this gene.

Gene
LRRC3C
Gene Name
leucine rich repeat containing 3C
There are no transcripts associated with this gene.

Gene
ORMDL3
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320801.1 377 Silent Mutation AGC,AGT S114S NP_001307730.1
NM_001320802.1 377 Silent Mutation AGC,AGT S114S NP_001307731.1
NM_001320803.1 377 Silent Mutation AGC,AGT S114S NP_001307732.1
NM_139280.3 377 Silent Mutation AGC,AGT S114S NP_644809.1
XM_017025345.1 377 Silent Mutation AGC,AGT S132S XP_016880834.1
XM_017025346.1 377 Silent Mutation AGC,AGT S114S XP_016880835.1
XM_017025347.1 377 Silent Mutation AGC,AGT S114S XP_016880836.1
XM_017025348.1 377 Silent Mutation AGC,AGT S114S XP_016880837.1

View Full Product Details