Product Details

SNP ID
rs143906788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7355225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCATGTGGAGGCTGGCACTAGG[C/T]GGGGTTTTCCTGGCAGCCGCCCAGG
Phenotype
MIM: 607763 MIM: 609848
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 112 Intron NP_055531.1
Gene
KCTD11
Gene Name
potassium channel tetramerization domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002914.2 112 Intron NP_001002914.1
Gene
TMEM95
Gene Name
transmembrane protein 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320435.1 112 Silent Mutation GGC,GGT G7G NP_001307364.1
NM_001320436.1 112 Silent Mutation GGC,GGT G7G NP_001307365.1
NM_198154.2 112 Silent Mutation GGC,GGT G7G NP_937797.1
XM_017024565.1 112 Silent Mutation GGC,GGT G7G XP_016880054.1
XM_017024566.1 112 Silent Mutation GGC,GGT G7G XP_016880055.1
XM_017024567.1 112 Silent Mutation GGC,GGT G7G XP_016880056.1
XM_017024568.1 112 Silent Mutation GGC,GGT G7G XP_016880057.1
XM_017024569.1 112 Silent Mutation GGC,GGT G7G XP_016880058.1
XM_017024570.1 112 Intron XP_016880059.1
XM_017024571.1 112 Silent Mutation GGC,GGT G7G XP_016880060.1

View Full Product Details