Product Details

SNP ID
rs143976560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10443396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTACTCTTCACTTATGACTTT[C/T]GTGTGAACCTCCCGACTCTTCACTC
Phenotype
MIM: 160742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYH4 PubMed Links

Gene Details

Gene
MYH4
Gene Name
myosin heavy chain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017533.2 6054 Silent Mutation ACA,ACG T1933T NP_060003.2
XM_017024676.1 6054 Silent Mutation ACA,ACG T1933T XP_016880165.1
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details