Product Details

SNP ID
rs144071223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38853761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTCGATTCAAAGGACACTGGG[A/T]TATGAACTTTAACGCCCCGCCCACC
Phenotype
MIM: 603662
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPL23 PubMed Links

Gene Details

Gene
RPL23
Gene Name
ribosomal protein L23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000978.3 Intron NP_000969.1
Gene
SNORA21
Gene Name
small nucleolar RNA, H/ACA box 21
There are no transcripts associated with this gene.

View Full Product Details