Product Details
- SNP ID
-
rs144150639
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:2337692 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGCCGCCTCCTGCCACACCATG[A/G]GCAGCGCAGAGGACGCAGTCAAAGA
- Phenotype
-
MIM: 611418
MIM: 611214
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SGSM2
PubMed Links
Gene Details
- Gene
- SGSM2
- Gene Name
- small G protein signaling modulator 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098509.1 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
NP_001091979.1 |
NM_014853.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
NP_055668.2 |
XM_011524101.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522403.1 |
XM_011524102.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522404.1 |
XM_011524103.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522405.1 |
XM_011524104.2 |
119 |
Intron |
|
|
XP_011522406.1 |
XM_011524105.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522407.1 |
XM_011524106.1 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522408.1 |
XM_011524107.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522409.1 |
XM_011524108.1 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522410.1 |
XM_011524109.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522411.1 |
XM_011524110.2 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_011522412.1 |
XM_017025474.1 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_016880963.1 |
XM_017025475.1 |
119 |
Missense Mutation |
AGC,GGC |
S2G |
XP_016880964.1 |
XM_017025476.1 |
119 |
Intron |
|
|
XP_016880965.1 |
- Gene
- SNORD91A
- Gene Name
- small nucleolar RNA, C/D box 91A
There are no transcripts associated with this gene.
- Gene
- SNORD91B
- Gene Name
- small nucleolar RNA, C/D box 91B
There are no transcripts associated with this gene.
- Gene
- TSR1
- Gene Name
- TSR1, ribosome maturation factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018128.4 |
119 |
Intron |
|
|
NP_060598.3 |
View Full Product Details