Product Details
- SNP ID
-
rs144257869
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:39766242 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGGAAGGTGGATGCTTTTCTTTT[C/T]CAGCTCTTGAGGGGCACCGTTTGAC
- Phenotype
-
MIM: 606221
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IKZF3
PubMed Links
Gene Details
- Gene
- IKZF3
- Gene Name
- IKAROS family zinc finger 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001257408.1 |
921 |
Missense Mutation |
AAA,GAA |
K326E |
NP_001244337.1 |
NM_001257409.1 |
921 |
Missense Mutation |
AAA,GAA |
K287E |
NP_001244338.1 |
NM_001257410.1 |
921 |
Missense Mutation |
AAA,GAA |
K273E |
NP_001244339.1 |
NM_001257411.1 |
921 |
Missense Mutation |
AAA,GAA |
K217E |
NP_001244340.1 |
NM_001257412.1 |
921 |
Missense Mutation |
AAA,GAA |
K178E |
NP_001244341.1 |
NM_001257413.1 |
921 |
Missense Mutation |
AAA,GAA |
K139E |
NP_001244342.1 |
NM_001257414.1 |
921 |
Missense Mutation |
AAA,GAA |
K226E |
NP_001244343.1 |
NM_001284514.1 |
921 |
Missense Mutation |
AAA,GAA |
K113E |
NP_001271443.1 |
NM_001284515.1 |
921 |
Missense Mutation |
AAA,GAA |
K113E |
NP_001271444.1 |
NM_001284516.1 |
921 |
Missense Mutation |
AAA,GAA |
K113E |
NP_001271445.1 |
NM_012481.4 |
921 |
Missense Mutation |
AAA,GAA |
K360E |
NP_036613.2 |
NM_183228.2 |
921 |
Missense Mutation |
AAA,GAA |
K304E |
NP_899051.1 |
NM_183229.2 |
921 |
Missense Mutation |
AAA,GAA |
K321E |
NP_899052.1 |
NM_183230.2 |
921 |
Missense Mutation |
AAA,GAA |
K321E |
NP_899053.1 |
NM_183231.2 |
921 |
Missense Mutation |
AAA,GAA |
K265E |
NP_899054.1 |
NM_183232.2 |
921 |
Missense Mutation |
AAA,GAA |
K282E |
NP_899055.1 |
View Full Product Details