Product Details

SNP ID
rs144269232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59210195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCATGGCGGGAGGATGAGATCTG[C/T]GTTGTGGGAATCTTCGGCAAGACGG
Phenotype
MIM: 615920 MIM: 613175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRR11 PubMed Links

Gene Details

Gene
PRR11
Gene Name
proline rich 11
There are no transcripts associated with this gene.

Gene
SMG8
Gene Name
SMG8, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018149.6 186 Silent Mutation TGC,TGT C48C NP_060619.4

View Full Product Details