Product Details

SNP ID
rs144282468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76314111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATACATGTTACTTTGAATTTTC[C/T]AGTAGCCACATTAAAAACAGATGAA
Phenotype
MIM: 601249
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPSAP1 PubMed Links

Gene Details

Gene
PRPSAP1
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002766.2 Intron NP_002757.2
XM_011525034.2 Intron XP_011523336.1
Gene
QRICH2
Gene Name
glutamine rich 2
There are no transcripts associated with this gene.

View Full Product Details