Product Details
- SNP ID
-
rs144304911
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:43753951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGAAGAATTGCCAAAAAAACGAC[C/G]AGTCCTGCAGGCTTTCATATGTCAT
- Phenotype
-
MIM: 605740
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SOST
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17885979] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SOST
- Gene Name
- sclerostin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025237.2 |
2080 |
UTR 3 |
|
|
NP_079513.1 |
View Full Product Details