Product Details
- SNP ID
-
rs144428416
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:49711422 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCCCGGGTTTGAGGGGCAGTTAC[A/G]GAAGATGAAGCCCATGCCGGGGAAG
- Phenotype
-
MIM: 610790
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM117A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34759387] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM117A
- Gene Name
- family with sequence similarity 117 member A
- Gene
- SLC35B1
- Gene Name
- solute carrier family 35 member B1
There are no transcripts associated with this gene.
View Full Product Details