Product Details

SNP ID
rs144531533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82322917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAACAGCGGCACCTGTACCAGGC[A/G]AACATGACCAGAGCGACCAAGAGGA
Phenotype
MIM: 186820 MIM: 602602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD7 PubMed Links

Gene Details

Gene
CD7
Gene Name
CD7 molecule
There are no transcripts associated with this gene.

Gene
SECTM1
Gene Name
secreted and transmembrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003004.2 927 Silent Mutation TTC,TTT F166F NP_002995.1
XM_005256392.3 927 Silent Mutation TTC,TTT F166F XP_005256449.1
XM_011523588.2 927 Silent Mutation TTC,TTT F166F XP_011521890.1

View Full Product Details